missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxysalicylate, 98%, Thermo Scientific™

Product Code. 10285890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285890 10 g Glass bottle 10g
10046393 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285890 Supplier Thermo Scientific Acros Supplier No. 174270100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5446-02-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008424
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Synonym Methyle2-hydroxy-4-methoxybenzoate
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C=C1O

Specifications

Melting Point 49°C to 53°C
Color White to Yellow
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)CO2CH3
Quantity 10 g
Beilstein 10, 381
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-methoxysalicylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.