missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorobenzoylacetate, 99%, Thermo Scientific™

Product Code. 10096093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096093 25 g Glass bottle 25g
10700711 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10096093 Supplier Thermo Scientific Acros Supplier No. 199760250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 63131-29-3
Molecular Formula C10H9FO3
Molecular Weight (g/mol) 196.18
MDL Number MFCD00000355
InChI Key HGLVYXXPRSNKQN-UHFFFAOYSA-N
Synonym methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6
PubChem CID 579425
IUPAC Name methyl 3-(4-fluorophenyl)-3-oxopropanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)F

Specifications

Color Colorless to Yellow
Density 1.2200g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4COCH2CO2CH3
Refractive Index 1.5200 to 1.522
Quantity 25 g
Specific Gravity 1.22
Formula Weight 196.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 4-fluorobenzoylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.