missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97%

Product Code. 10236910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10236910 25 mL 25mL
10297543 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236910 Supplier Thermo Scientific Acros Supplier No. 241620250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32807-28-6
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000938
InChI Key HFLMYYLFSNEOOT-UHFFFAOYSA-N
Synonym methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
PubChem CID 36240
IUPAC Name methyl 4-chloro-3-oxobutanoate
SMILES COC(=O)CC(=O)CCl

Specifications

Melting Point 14.0°C to 16.0°C
Color Orange-Yellow to Yellow
Density 1.2800g/mL
Boiling Point 212.0°C to 213.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C(O)CH2COOCH3
Refractive Index 1.4560 to 1.4580
Quantity 25 mL
Specific Gravity 1.28
Solubility Information Solubility in water: 71g/L (20 c)
Formula Weight 150.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 4-chloroacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.