missing translation for 'onlineSavingsMsg'
Learn More

Methyle4-aminotetrahydropyran-4-carboxylate hydrochloride, 95%, Maybridge

Product Code. 10412284
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10412284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10412284 Supplier Thermo Scientific Maybridge Supplier No. MO08533DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 184698-41-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H13NO3
MDL Number MFCD09608001
Quantity 1 g
Synonym methyl 4-aminotetrahydro-2h-pyran-4-carboxylate, methyl 4-aminotetrahydropyran-4-carboxylate, 4-aminotetrahydropyran-4-carboxylic acid methyl ester, 2h-pyran-4-carboxylic acid, 4-aminotetrahydro-, methyl ester, methyl 4-amino-tetrahydro-2h-pyran-4-carboxylate, 4-amino-4-methoxycarbonyl-tetrahydropyrane, 2h-pyran-4-carboxylicacid, 4-aminotetrahydro-, methyl ester, 2h-pyran-4-carboxylic acid, 4-aminotetrahydro-, methyl ester 9ci, methyl 4-aminotetrahydropyran-4-carboxylate hydrochloride;2h-pyran-4-carboxylicacid,4-aminotetrahydro-,methylester 9ci
InChI Key AZFBPYNPITWILD-UHFFFAOYSA-N
SMILES COC(=O)C1(N)CCOCC1
IUPAC Name methyl 4-aminooxane-4-carboxylate
Molecular Weight (g/mol) 159.19
PubChem CID 18698314
Formula Weight 195.64
Percent Purity 97%
Chemical Name or Material Methyl 4-aminotetrahydropyran-4-carboxylate hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.