missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(aminomethyl)benzoate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 10254954
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6232-11-7
C9H11NO2·ClH
201.65
MFCD00182671
GIZCKBSSWNIUMZ-UHFFFAOYSA-N
methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride
2729253
methyl 4-(aminomethyl)benzoate;hydrochloride
COC(=O)C1=CC=C(C=C1)CN.Cl
This item is not returnable. View return policy
6232-11-7
C9H11NO2·ClH
201.65
MFCD00182671
GIZCKBSSWNIUMZ-UHFFFAOYSA-N
methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride
2729253
methyl 4-(aminomethyl)benzoate;hydrochloride
COC(=O)C1=CC=C(C=C1)CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 234.5°C to 237.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Linear Formula NH2CH2C6H4COOCH3
Quantity 25 g
Formula Weight 201.65
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(aminomethyl)benzoate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-(aminomethyl)benzoate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'