missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 99%, Thermo Scientific™

Product Code. 10330744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330744 5 g Glass bottle 5g
10562071 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10330744 Supplier Thermo Scientific Acros Supplier No. 292780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 159660-85-2
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00216930
InChI Key HLKWMBMESNOAMS-UHFFFAOYSA-N
Synonym methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester
PubChem CID 563385
IUPAC Name methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate
SMILES COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N

Specifications

Melting Point 87.0°C to 89.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 246.31
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.