missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrobenzoate, 98%

Product Code. p-3758430 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10783393

Brand: Thermo Scientific Acros 168440250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-95-1 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 76°C to 80°C
Color Beige
Boiling Point 279°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CO2CH3
Quantity 25 g
Beilstein 09, 378
Merck Index 10, 6435
Solubility Information Solubility in water: insoluble
Formula Weight 181.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-nitrobenzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.