missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-methyl-4-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 10270495
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
Unit Size:
100g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10270495 100 g 100g
10203572 1 g 1g
10579972 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10270495 Supplier Thermo Scientific Maybridge Supplier No. TL00715FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24078-21-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00085640
InChI Key IEFONJKJLZFGKQ-UHFFFAOYSA-N
Synonym benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
PubChem CID 260927
IUPAC Name methyl 3-methyl-4-nitrobenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 100 g
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.