missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10426292
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10426292 50 g Amber Glass Bottle 50g
10263232 10 g Amber Glass Bottle 10g
10397584 100 g Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10426292 Supplier Thermo Scientific Maybridge Supplier No. SB01952EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C6H6O3S
MDL Number MFCD00055642

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyle3-hydroxythiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.