Learn More
Thermo Scientific Maybridge Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate, 10g, 97%
Brand: Thermo Scientific Maybridge BTB04875EA
Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate, 10g, 97%
Code : 31
Additional Details : CAS Number : 13134-76-4 Weight : 0.06000kg