missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorophenylacetate, 98%, Thermo Scientific™

Product Code. 10005163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10005163 100 g Glass bottle 100g
10425243 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10005163 Supplier Thermo Scientific Acros Supplier No. 371671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 53088-68-9
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00800679
InChI Key HTBBVKFFDDQVER-UHFFFAOYSA-N
PubChem CID 2733544
IUPAC Name methyl 2-(3-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(Cl)=C1

Specifications

Color Colorless to Yellow
Density 1.2000g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5225 to 1.5245
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.2
Linear Formula ClC6H4CH2C(O)OCH3
Synonym methyl 2-3-chlorophenyl acetate, methyl 3-chlorophenylacetate, methyl 3-chlorophenyl acetate, phenylacetic acid, 3-chloro, methyl ester, benzeneacetic acid, 3-chloro-, methyl ester, methyl2-3-chlorophenyl acetate, methyl 2-3-chlorophenyl ethanoate, 3-chlorophenylacetic acid methyl ester, 3-chlorophenyl acetic acid methyl ester
Formula Weight 184.51
Percent Purity 98%
Chemical Name or Material Methyl 3-chlorophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-chlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.