missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorobenzoate, 98%, Acros Organics™

Product Code. 10124700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
Glass bottle; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10124700 Glass bottle; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124700 Supplier Thermo Scientific Acros Supplier No. 215590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2905-65-9
Melting Point 21.0°C
Color Colorless to Yellow
Density 104g/mL
Boiling Point 231.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H7ClO2
Linear Formula ClC6H4CO2CH3
Refractive Index 1.5310 to 1.5330
MDL Number MFCD00000592
Quantity 25 g
Beilstein 09, 338
Merck Index 10, 2093
Synonym methyl m-chlorobenzoate, benzoic acid, 3-chloro-, methyl ester, m-chlorobenzoic acid methyl ester, methyl-3-chlorobenzoate, benzoic acid, m-chloro-, methyl ester, 3-chlorobenzoic acid methyl ester, 3-chloro-benzoic acid methyl ester, acmc-1cegg, dsstox_cid_7507, 3-chlorobenzoic acid methyl
InChI Key XRDRKVPNHIWTBX-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC(Cl)=C1
IUPAC Name methyl 3-chlorobenzoate
Molecular Weight (g/mol) 170.59
PubChem CID 17946
Formula Weight 170.6
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl 3-chlorobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-chlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.