Learn More
Methyle3-chloro-4-methylthiophene-2-carboxylate, 97%, Thermo Scientific™
£13.80 - £71.35
Chemical Identifiers
CAS | 175137-11-8 |
---|---|
Molecular Formula | C7H7ClO2S |
Molecular Weight (g/mol) | 190.641 |
MDL Number | MFCD00052072 |
InChI Key | BUFSIDWTJVUAER-UHFFFAOYSA-N |
Synonym | methyl 3-chloro-4-methyl-2-thiophenecarboxylate, 3-chloro-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-chloro-4-methyl-, methyl ester, maybridge1_003874, 2-methoxycarbonyl-3-chloro4-methylthiophene, 2-methoxycarbonyl-3-chloro-4-methylthiophene, methyl 3-chloro-4-methylthiophenecarboxylate, methyl3-chloro-4-methylthiophene-2-carboxylate, methyl 3-chloranyl-4-methyl-thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-chloro-4-methyl-, methyl ester |
PubChem CID | 2777567 |
IUPAC Name | methyl 3-chloro-4-methylthiophene-2-carboxylate |
SMILES | CC1=CSC(=C1Cl)C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10150592
|
Thermo Scientific Maybridge
GK01297DA |
1 g | Amber Glass Bottle |
£13.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10069411
|
Thermo Scientific Maybridge
GK01297EA |
10 g | Amber Glass Bottle |
£71.35
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175137-11-8 | |
190.641 | |
BUFSIDWTJVUAER-UHFFFAOYSA-N | |
2777567 | |
CC1=CSC(=C1Cl)C(=O)OC |
C7H7ClO2S | |
MFCD00052072 | |
methyl 3-chloro-4-methyl-2-thiophenecarboxylate, 3-chloro-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-chloro-4-methyl-, methyl ester, maybridge1_003874, 2-methoxycarbonyl-3-chloro4-methylthiophene, 2-methoxycarbonyl-3-chloro-4-methylthiophene, methyl 3-chloro-4-methylthiophenecarboxylate, methyl3-chloro-4-methylthiophene-2-carboxylate, methyl 3-chloranyl-4-methyl-thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-chloro-4-methyl-, methyl ester | |
methyl 3-chloro-4-methylthiophene-2-carboxylate |
Specifications
175137-11-8 | |
100.0 | |
C7H7ClO2S | |
methyl 3-chloro-4-methyl-2-thiophenecarboxylate, 3-chloro-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-chloro-4-methyl-, methyl ester, maybridge1_003874, 2-methoxycarbonyl-3-chloro4-methylthiophene, 2-methoxycarbonyl-3-chloro-4-methylthiophene, methyl 3-chloro-4-methylthiophenecarboxylate, methyl3-chloro-4-methylthiophene-2-carboxylate, methyl 3-chloranyl-4-methyl-thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-chloro-4-methyl-, methyl ester | |
CC1=CSC(=C1Cl)C(=O)OC | |
190.641 | |
190.65 | |
Methyl 3-chloro-4-methylthiophene-2-carboxylate |
96.5 | |
97% | |
MFCD00052072 | |
BUFSIDWTJVUAER-UHFFFAOYSA-N | |
methyl 3-chloro-4-methylthiophene-2-carboxylate | |
2777567 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.