missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(bromomethyl)benzoate, 95%

Product Code. 10339143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339143 100 g Glass bottle 100g
10500552 5 g Glass bottle 5g
10242622 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10339143 Supplier Thermo Scientific Acros Supplier No. 368631000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1129-28-8 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1

Specifications

Melting Point 35.0°C to 37.0°C
Color White
Boiling Point 135.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH2BrC6H4C(O)OCH3
Quantity 100 g
Beilstein 09, IV, 1721
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 229
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 3-(bromomethyl)benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-(bromomethyl)benzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.