missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10245524
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10245524

Brand: Thermo Scientific Maybridge GK00598EB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91076-93-6
Molecular Formula C12H10ClNO2S
Molecular Weight (g/mol) 267.727
MDL Number MFCD00052591
InChI Key MELGGDOYSMRBGA-UHFFFAOYSA-N
PubChem CID 2777492
IUPAC Name methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=C(C=C2)Cl)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Synonym methyl 3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl-3-amino-5-4-chlorophenyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-amino-5-4-chlorophenyl-, methyl ester, 3-amino-5-4-chlorophenyl thiophene-2-carboxylic acid methyl ester, pubchem8387, maybridge1_003891, methyl 3-amino-5-4-chlorophenyl thiophene-2carboxylate, methyl 3-amino-5-4-chlorophenyl-2-thiophenecarboxylate, methyl 3-amino-5-4-chlorophenyl-thiophene-2-carboxylate
Formula Weight 267.74
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.