missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10324382
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10324382 10 g Amber Glass Bottle 10g
10364332 1 g Amber Glass Bottle 1g
10245524 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10324382 Supplier Thermo Scientific Maybridge Supplier No. GK00598EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91076-93-6
Molecular Formula C12H10ClNO2S
Molecular Weight (g/mol) 267.727
MDL Number MFCD00052591
InChI Key MELGGDOYSMRBGA-UHFFFAOYSA-N
PubChem CID 2777492
IUPAC Name methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=C(C=C2)Cl)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym methyl 3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl-3-amino-5-4-chlorophenyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-amino-5-4-chlorophenyl-, methyl ester, 3-amino-5-4-chlorophenyl thiophene-2-carboxylic acid methyl ester, pubchem8387, maybridge1_003891, methyl 3-amino-5-4-chlorophenyl thiophene-2carboxylate, methyl 3-amino-5-4-chlorophenyl-2-thiophenecarboxylate, methyl 3-amino-5-4-chlorophenyl-thiophene-2-carboxylate
Formula Weight 267.74
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.