missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | methyl 3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl-3-amino-5-4-chlorophenyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-amino-5-4-chlorophenyl-, methyl ester, 3-amino-5-4-chlorophenyl thiophene-2-carboxylic acid methyl ester, pubchem8387, maybridge1_003891, methyl 3-amino-5-4-chlorophenyl thiophene-2carboxylate, methyl 3-amino-5-4-chlorophenyl-2-thiophenecarboxylate, methyl 3-amino-5-4-chlorophenyl-thiophene-2-carboxylate |
Formula Weight | 267.74 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'