missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10364332
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 91076-93-6
Molecular Formula C12H10ClNO2S
Molecular Weight (g/mol) 267.727
MDL Number MFCD00052591
InChI Key MELGGDOYSMRBGA-UHFFFAOYSA-N
PubChem CID 2777492
IUPAC Name methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=C(C=C2)Cl)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym methyl 3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl-3-amino-5-4-chlorophenyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-amino-5-4-chlorophenyl-, methyl ester, 3-amino-5-4-chlorophenyl thiophene-2-carboxylic acid methyl ester, pubchem8387, maybridge1_003891, methyl 3-amino-5-4-chlorophenyl thiophene-2carboxylate, methyl 3-amino-5-4-chlorophenyl-2-thiophenecarboxylate, methyl 3-amino-5-4-chlorophenyl-thiophene-2-carboxylate
Formula Weight 267.74
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'