missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10376704
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10376704 25 g 25g
10651293 1 g 1g
10304722 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376704 Supplier Thermo Scientific Maybridge Supplier No. KM01952EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175201-72-6
Molecular Formula C9H13NO4S2
Molecular Weight (g/mol) 263.33
MDL Number MFCD00067930
InChI Key LSLWECVRXQXIRH-UHFFFAOYSA-N
Synonym methyl 3-amino-4-isopropylsulfonyl thiophene-2-carboxylate, methyl 3-amino-4-propane-2-sulfonyl thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-1-methylethyl sulfonyl-, methyl ester, methyl 3-amino-4-methylethyl sulfonyl thiophene-2-carboxylate, maybridge1_004550, methyl 3-amino-4-isopropylsulfonyl thiophene-2carboxylate, methyl 3-azanyl-4-propan-2-ylsulfonyl-thiophene-2-carboxylate, 3-amino-4-propane-2-sulfonyl-thiophene-2-carboxylic acid methyl ester, 3-amino-4-propan-2-ylsulfonyl-2-thiophenecarboxylic acid methyl ester
PubChem CID 2778568
IUPAC Name methyl 3-amino-4-propan-2-ylsulfonylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(=CS1)S(=O)(=O)C(C)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 263.34
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.