missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle3-amino-4-cyanothiophene-2-carboxylate, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 10g
Specifications
CAS | 102123-28-4 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C7H6N2O2S |
MDL Number | MFCD00052590 |
Quantity | 10 g |
Synonym | 2-thiophenecarboxylicacid, 3-amino-4-cyano-, methyl ester, acmc-1boui, methyl 3-amino-4-cyano thiophene-2-carboxylate, methyl-3-amino-4-cyanothiophene-2-carboxylate, 2-thiophenecarboxylicacid,3-amino-4-cyano-,methyl ester, 3-amino-4-cyanothiophene-2-carboxylic acid methyl ester |
InChI Key | AEWVEROSMGRHRG-UHFFFAOYSA-N |
SMILES | COC(=O)C1=C(C(=CS1)C#N)N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'