missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-thiophenecarboxylate, 99%

Product Code. 10378680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10378680 50 g Glass bottle 50g
10368680 10 g Glass bottle 10g
10398703 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10378680 Supplier Thermo Scientific Acros Supplier No. 214300500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22288-78-4 | C6H7NO2S | 157.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22288-78-4
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00009765
InChI Key TWEQNZZOOFKOER-UHFFFAOYSA-N
Synonym methyl 3-amino-2-thiophenecarboxylate, 3-aminothiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-, methyl ester, 3-amino-thiophene-2-carboxylic acid methyl ester, 3-amino-2-thiophenecarboxylic acid methyl ester, methyl 3-amino-2-thiophene carboxylate, methyl-3-amino-thiophene-2-carboxylate, 3-amino-2-carbomethoxythiophene, 3-amino-2-methoxycarbonylthiophene, methyl3-aminothiophene-2-carboxylate
PubChem CID 89652
IUPAC Name methyl 3-aminothiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C=CS1

Specifications

Melting Point 66°C to 67°C
Color Beige to Beige-Brown
Boiling Point 100°C to 102°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Formula Weight 157.19
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Methyl 3-amino-2-thiophenecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-amino-2-thiophenecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.