missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-pyrazinecarboxylate, 98%, Acros Organics™

Brand:  Acros Organics 269590500

Code : 29

Additional Details : CAS Number : 16298-03-6 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10195833

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Methyl 3-amino-2-pyrazinecarboxylate
16298-03-6
98%
153.14
98%
≥97.5%
C6H7N3O2
MFCD00010097

Specifications

Methyl 3-amino-2-pyrazinecarboxylate
166° to 172°C
98%
Authentic
Crystalline Powder
98%
153.14
25, IV, 4412
Yellow
50g
16298-03-6
Glass bottle
≥97.5%
C6H7N3O2
MFCD00010097
Videos
SDS