missing translation for 'onlineSavingsMsg'
Learn More

Methyle3,4-diaminobenzoate, 97%, Thermo Scientific™

Product Code. 10387294
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387294 25 g Amber Glass Bottle 25g
10570742 1 g Amber Glass Bottle 1g
10713063 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387294 Supplier Thermo Scientific Maybridge Supplier No. RDP00052EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 36692-49-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017098
InChI Key IOPLHGOSNCJOOO-UHFFFAOYSA-N
Synonym 3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate
PubChem CID 135524
IUPAC Name methyl 3,4-diaminobenzoate
SMILES COC(=O)C1=CC=C(N)C(N)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3OC(O)C6H3(NH2)2
Quantity 25 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Methyl 3, 4-diaminobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.