missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trihydroxybenzoate, 99%

Product Code. 10440701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10440701 50 g Plastic bottle 50g
10760741 5 g Glass bottle 5g
10616924 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10440701 Supplier Thermo Scientific Acros Supplier No. 243460500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-24-1 | C8H8O5 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-24-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.15
MDL Number MFCD00002194
InChI Key FBSFWRHWHYMIOG-UHFFFAOYSA-N
Synonym methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
IUPAC Name methyl 3,4,5-trihydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 199°C to 203°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 4375
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol,methanol,ether
Formula Weight 184.15
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4,5-trihydroxybenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3,4,5-trihydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.