missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-chloro-6-methylpyrimidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10427252
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10427252 1 g Amber Glass Bottle 1g
10111042 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10427252 Supplier Thermo Scientific Maybridge Supplier No. SPB06334DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89793-11-3
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD00117906
InChI Key ZNPORLVPCMFKOR-UHFFFAOYSA-N
Synonym methyl2-chloro-6-methylpyrimidine-4-carboxylate, methyl 2-chloro-4-methylpyrimidine-6-carboxylate, methyl 2-chloro-6-methyl-pyrimidine-4-carboxylate, 4-pyrimidinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyrimidine, 2-chloro-6-methyl-pyrimidine-4-carboxylic acid methyl ester, acmc-20akio, 2-chloro-4-carbomethoxy-6-methylpyrimidine, methyl 2-chloro-6-methylpyrimidine 4-carboxylate, methyl-2-chloro-6-methylpyrimidine-4-carboxylate
PubChem CID 2736835
IUPAC Name methyl 2-chloro-6-methylpyrimidine-4-carboxylate
SMILES CC1=NC(=NC(=C1)C(=O)OC)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material Methyle2-chloro-6-methylpyrimidine-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.