missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-amino-4,5-dimethoxybenzoate, 97%, Thermo Scientific™

Product Code. 10722693
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722693 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10722693 Supplier Thermo Scientific Maybridge Supplier No. SB01989DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 26759-46-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H13NO4
MDL Number MFCD00014904
Quantity 1 g
Synonym methyl 6-aminoveratrate, methyl 4,5-dimethoxyanthranilate, methyl 2-amino-4,5-dimethoxy benzoate, 2-amino-4,5-dimethoxybenzoic acid methyl ester, 6-aminoveratric acid methyl ester, methyl2-amino-4,5-dimethoxybenzoate, 4,5-dimethoxyanthranilic acid methyl, benzoic acid, 2-amino-4,5-dimethoxy-, methyl ester, methyl-2-amino-4,5-dimethoxybenzoate, 2-amino-4,5-dimethoxy-benzoic acid methyl ester
InChI Key QQFHCCQSCQBKBG-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(OC)=C(OC)C=C1N
IUPAC Name methyl 2-amino-4,5-dimethoxybenzoate
Molecular Weight (g/mol) 211.22
PubChem CID 611144
Formula Weight 211.22
Percent Purity 97%
Chemical Name or Material Methyl 2-amino-4, 5-dimethoxybenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.