missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate, 97%, Maybridge™

Product Code. p-2704842
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10039551 1 g 1g
10564885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10039551 Supplier Thermo Scientific Maybridge Supplier No. RJC01802DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 114524-80-0
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.283
MDL Number MFCD00152138
InChI Key RCUNGDZWHFRBBP-UHFFFAOYSA-N
Synonym methyl 2-amino-3-1h-indol-3-yl-2-methylpropanoate, alpha-methyl-dl-tryptophan methyl ester, tryptophan,, a-methyl-, methyl ester, alpha-methyl-d,l-tryptophan methyl ester, methyl 2-azanyl-3-1h-indol-3-yl-2-methyl-propanoate, maybridge1_006684, acmc-20ekci, pubchem14800, h-me-dl-trp-ome, h-2-me-dl-try-ome
PubChem CID 2795140
IUPAC Name methyl 2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
SMILES CC(CC1=CNC2=CC=CC=C21)(C(=O)OC)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.