missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Thermo Scientific™

Product Code. 10814151
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 72784-42-0
Molecular Formula C5H9NO2·ClH
Molecular Weight (g/mol) 151.59
InChI Key CHSDCWKLSHPKFY-UHFFFAOYSA-N
Synonym methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, methyl 1-aminocyclopropane-1-carboxylate hcl, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropanecarboxylate hcl, methyl-1-aminocyclopropanecarboxylate hydrochloride
PubChem CID 12760695
IUPAC Name methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES COC(=O)C1(CC1)N.Cl

Specifications

Melting Point 176.4°C to 178.7°C
Color White
Elemental Analysis 38.37 to 40.33% C, 8.95 to 9.40%
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 500 mg
Formula Weight 151.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-aminocyclopropanecarboxylate hydrochloride
compliance-icons
Product Identifier
  • Methyl 1-aminocyclopropanecarboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'