missing translation for 'onlineSavingsMsg'
Learn More

Methyle1,2,3-benzothiadiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2704525
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
23616-15-1
C8H6N2O2S
194.208
MFCD01566550
JCRFJMHWEAYQAA-UHFFFAOYSA-N
methyl benzo d 1,2,3 thiadiazole-5-carboxylate, methyl1,2,3-benzothiadiazole-5-carboxylate, 5-carbomethoxy-1,2,3-benzothiadiazole, maybridge1_005151, 5-methoxycarbonyl-1,2,3-benzothiadiazole, methyl benzo 1,2,3 thiadiazole-5-carboxylate, methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 4-methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 1,2,3-benzothiadiazole-5-carboxylic acid methyl ester, 1,2,3-benzothiadiazole-5-carboxylicacid, methyl ester
2736813
methyl 1,2,3-benzothiadiazole-5-carboxylate
COC(=O)C1=CC2=C(C=C1)SN=N2
This item is not returnable. View return policy
23616-15-1
C8H6N2O2S
194.208
MFCD01566550
JCRFJMHWEAYQAA-UHFFFAOYSA-N
methyl benzo d 1,2,3 thiadiazole-5-carboxylate, methyl1,2,3-benzothiadiazole-5-carboxylate, 5-carbomethoxy-1,2,3-benzothiadiazole, maybridge1_005151, 5-methoxycarbonyl-1,2,3-benzothiadiazole, methyl benzo 1,2,3 thiadiazole-5-carboxylate, methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 4-methylbenzo d 1,2,3 thiadiazole-5-carboxylate, 1,2,3-benzothiadiazole-5-carboxylic acid methyl ester, 1,2,3-benzothiadiazole-5-carboxylicacid, methyl ester
2736813
methyl 1,2,3-benzothiadiazole-5-carboxylate
COC(=O)C1=CC2=C(C=C1)SN=N2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.21
Percent Purity 97%
Chemical Name or Material Methyle1, 2, 3-benzothiadiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'