missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyldopa hydrate, MedChemExpress
Description
Methyldopa hydrate (L-(-)-α-Methyldopa hydrate), a potent antihyoertensive agent, is an alpha-adrenergic agonist (selective for α2-adrenergic receptors). Methyldopa hydrate is a prodrug and is metabolized (α-Methylepinephrine) in the central nervous system.
Specifications
Specifications
| Chemical Name or Material | Methyldopa hydrate |
| CAS | 41372-08-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H16NO5.5 |
| Quantity | 1 g |
| Synonym | L-(-)-α-Methyldopa hydrateMK-351 hydrate |
| Solubility Information | DMSO : 33.33 mg/mL (139.90 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.20 mM; Need ultrasonic) |
| SMILES | OC1=C(O)C=CC(C[C@@](C)(C(O)=O)N)=C1.[1.5H2O] |
| Molecular Weight (g/mol) | 238.24 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?