missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog Number p-3779101
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1486-28-8
C13H13P
200.22
MFCD00008508
UJNZOIKQAUQOCN-UHFFFAOYSA-N
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
73879
methyl(diphenyl)phosphane
CP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1486-28-8
C13H13P
200.22
MFCD00008508
UJNZOIKQAUQOCN-UHFFFAOYSA-N
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
73879
methyl(diphenyl)phosphane
CP(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 118.0°C to 120.0°C (2.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2PCH3
Refractive Index 1.6240 to 1.6260
Quantity 5 g
Beilstein 16, II, 371
Specific Gravity 1.076
Solubility Information Solubility in water: insoluble
Formula Weight 200.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyldiphenylphosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'