missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals

Catalog Number p-7029668
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
1486-28-8
C13H13P
200.22
MFCD00008508
UJNZOIKQAUQOCN-UHFFFAOYSA-N
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
73879
methyl(diphenyl)phosphane
CP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1486-28-8
C13H13P
200.22
MFCD00008508
UJNZOIKQAUQOCN-UHFFFAOYSA-N
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
73879
methyl(diphenyl)phosphane
CP(C1=CC=CC=C1)C1=CC=CC=C1

Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.064
Boiling Point 160°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.21
Percent Purity 99%
Chemical Name or Material Methyldiphenylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'