missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadienylmanganese tricarbonyl, 97%

Product Code. 11448630
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448630 1 g 1g
11458630 5 g 5g
11468630 25 g 25g
11438630 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448630 Supplier Thermo Scientific Alfa Aesar Supplier No. 047125.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12108-13-3 | C9H7MnO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12108-13-3
Molecular Formula C9H7MnO3
MDL Number MFCD00001426
Synonym MMT

Specifications

Melting Point 1.5°C
Color Yellow
Density 1.388 g/mL
Boiling Point 233°C
Flash Point 96°C (205°F)
Refractive Index 1.5873
Quantity 1 g
UN Number UN3281
Sensitivity Air sensitive
Merck Index 14,6223
Solubility Information Insoluble in water; Soluble in hydrocarbon solvents, alcohols, ether, acetone
Formula Weight 218.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.