missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadiene dimer, tech. 90%, stab. with 200ppm 4-tert-butylcatechol

Product Code. 11408227
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5000 mL
Unit Size:
1000mL
5000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408227 5000 mL 5000mL
11488790 500 mL 500mL
11498217 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408227 Supplier Thermo Scientific Alfa Aesar Supplier No. A17596.0J
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26472-00-4 | C12H16

Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26472-00-4
Molecular Formula C12H16
MDL Number MFCD00082297

Specifications

Melting Point ∼-50°C
Density 0.94
Boiling Point 198°C to 201°C
Flash Point 26°C (79°F)
Refractive Index 1.498
Quantity 5000 mL
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 160.26
Percent Purity 90%
Grade Technical
Chemical Name or Material Methylcyclopentadiene dimer, stabilized with 200ppm 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.