missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadiene dimer, 90%, stabilized

Product Code. 10347810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347810 250 g Glass bottle 250g
10031720 1 kg Glass bottle 1kg
10403405 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10347810 Supplier Thermo Scientific Acros Supplier No. 156852500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26472-00-4 | C12H16 | 160.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26472-00-4
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00082297

Specifications

Melting Point -51.0°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 200.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.4975 to 1.4995
Quantity 250 g
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene and ether
Formula Weight 160.26
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methylcyclopentadiene dimer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methylcyclopentadiene dimer
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.