missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclohexane-d14 (Reagent grade), CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Methylcyclohexane-D14 |
| CAS | 10120-28-2 |
| Molecular Formula | C7D14 |
| Quantity | 5 g |
| Synonym | Cyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11, 6-(methyl-d3)- (ACI), 6-(Methyl-d3)cyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11 (ACI), Cyclohexane-d11, methyl-d3- (7 CI, 8 CI, 9 CI), Methylcyclohexane-d14, Methylcyclohexane-D14 |
| InChI Formula | InChI=1 S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7 D |
| SMILES | [2 H]C1(C(C(C(C(C1([2 H])[2 H])([2 H])[2 H])([2 H])C([2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6-undecadeuterio-6-(trideuteriomethyl)cyclohexane |
| Molecular Weight (g/mol) | 112.27 |
| Formula Weight | 112.1974 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction