missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99+%

Product Code. p-7049948
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11358297

Brand: Thermo Scientific Alfa Aesar 031861.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-87-2 | C7H14 | 98.19 g/mol

Methylcyclohexane is used as rubber, paint and varnish, used solvents, oils, such as solvent extraction, which can also be used for organic synthesis, Standards for the calibration of the thermometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Color Colorless
Density 0.771 g/mL
Boiling Point 101°C
Flash Point -4°C (-25°F)
Refractive Index 1.423
Quantity 100 g
UN Number UN2296
Beilstein 505972
Merck Index 14,6047
Solubility Information Soluble in water 0.1g/L (20°C).
Formula Weight 98.19
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Methylcyclohexane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.