missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99+%, Thermo Scientific Chemicals

Catalog Number 11358297
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1

Methylcyclohexane is used as rubber, paint and varnish, used solvents, oils, such as solvent extraction, which can also be used for organic synthesis, Standards for the calibration of the thermometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -126°C
Color Colorless
Density 0.771 g/mL
Boiling Point 101°C
Flash Point -4°C (-25°F)
Refractive Index 1.423
Quantity 100 g
UN Number UN2296
Beilstein 505972
Merck Index 14,6047
Solubility Information Soluble in water 0.1g/L (20°C).
Formula Weight 98.19
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Methylcyclohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'