missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 98+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 13172024
31.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -126.3°C
Color Colorless
Density 0.7700g/mL
Boiling Point 101.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging AcroSeal glass Bottle
Linear Formula C6H11CH3
Refractive Index 1.4210 to 1.4230
Quantity 100 mL
Residue after Evaporation 0.001% max.
Merck Index 15, 6118
Specific Gravity 0.77
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
Formula Weight 98.19
Percent Purity 98+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Methylcyclohexane, Extra Dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'