missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylbenactyzium Bromide, MedChemExpress
Description
Methylbenactyzium Bromide is a muscarinic acetylcholine receptor (mAChR) inhibitor.
Specifications
Specifications
| Chemical Name or Material | Methylbenactyzium Bromide |
| CAS | 3166-62-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H28BrNO3 |
| Quantity | 200 mg |
| Solubility Information | DMSO : 6 mg/mL (14.21 mM; Need ultrasonic and warming) |
| SMILES | O=C(OCC[N+](CC)(CC)C)C(C1=CC=CC=C1)(O)C2=CC=CC=C2.[Br-] |
| Molecular Weight (g/mol) | 422.36 |
| Formula Weight | 422.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?