missing translation for 'onlineSavingsMsg'
Learn More

Methylated Spirit (IDA 99), 99% (v/v), Pure, (Industrial Methylated Spirit, 74 0.P.)

Product Code. 11412884 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11412884 2.5 L Plastic Coated Glass bottle 2.5 L
10552904 1 L Amber glass bottle 1 L
11482874 2.5 L Amber glass bottle 2.5 L
11492874 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 11412884 Supplier Fisher Chemical Supplier No. M/4450/PB17
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

MDL Number 3568

Specifications

Melting Point -114.1°C
Color Colorless
Boiling Point 78.2°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.