missing translation for 'onlineSavingsMsg'
Learn More

Methylated Spirit (IDA 94), 94% (v/v), Pure, (Industrial Methylated Spirit 64 0.P.), Fisher Chemical™

Product Code. 15950957 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
5 L
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
Metal Can
Plastic Coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15950957

Brand: Fisher Chemical M/4400/21

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

MDL Number 3568

Specifications

Melting Point -114°C
Color Colorless
Boiling Point 78.2°C
Packaging HDPE Plastic Bottle
Quantity 5 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylated spirit industrial 64 O.P.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.