missing translation for 'onlineSavingsMsg'
Learn More

Methylated Spirit (IDA 94), 94% (v/v), Pure, (Industrial Methylated Spirit 64 0.P.), Fisher Chemical™

Product Code. 11462874 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
5 L
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
Metal Can
Plastic Coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11462874 2.5 L Plastic Coated Glass Bottle 2.5 L
15940957 1 L Amber Glass Bottle 1 L
11442874 2.5 L Amber Glass Bottle 2.5 L
11472874 2.5 L HDPE Plastic Bottle 2.5 L
15950957 5 L HDPE Plastic Bottle 5 L
11452874 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Product Code. 11462874 Supplier Fisher Chemical Supplier No. M/4400/PB17
  • ONLINE EXCLUSIVE PRICE
    £110.27
    £79.50 / 2.5 L
    Save £30.77
    27% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Chemical Identifiers

MDL Number 3568

Specifications

Melting Point -114°C
Color Colorless
Boiling Point 78.2°C
Packaging Plastic Coated Glass Bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylated spirit industrial
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl alcohol, Methyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.