missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylated Spirit (IDA 94), 94% (v/v), Pure, (Industrial Methylated Spirit 64 0.P.), Fisher Chemical™
579.69 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
5 L
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
Metal Can
Plastic Coated Glass Bottle
Specifications
Melting Point | -114°C |
Color | Colorless |
Boiling Point | 78.2°C |
Packaging | Metal Can |
Quantity | 25 L |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Methylated spirit industrial
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
- Acute toxicity Category 4
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'