Learn More
Methylated Lysine Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577770
Additional Details : Weight : 0.01000kg
Description
0.2-0.5 µg/mL of PA5-77770 was sufficient for detection of the methylated histone by western blot analysis using melanoma cells in TBSt. |Detects proteins containing methylated lysine residues.
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.Specifications
Methylated Lysine | |
Polyclonal | |
Unconjugated | |
Dimethylysine; Methyl lysine; Trimethyl lysine | |
Methylated KLH Conjugated | |
100 μg | |
Primary | |
-20°C | |
Liquid | |
Chemical |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and no preservative; pH 7.4 | |
Rabbit | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG |