missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylated Lysine Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5115718
Additional Details : Weight : 0.01000kg
Description
Pan Methylated Lysine Rabbit pAbdetects endogenous levels of total Pan Methylated Lysine Rabbit pAb.
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.Specifications
Methylated Lysine | |
Polyclonal | |
Unconjugated | |
Dimethylysine; Methyl lysine; Trimethyl lysine | |
A synthesized peptide derived from human Pan Methylated Lysine, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
-20°C | |
Liquid | |
Chemical |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG |