missing translation for 'onlineSavingsMsg'
Learn More

Methylamine, extra pure, 40 wt% solution in water

Product Code. 10592901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10592901 2.5 L 2.5 L
10687282 1 L 1 L
10782564 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10592901 Supplier Thermo Scientific Acros Supplier No. 126230025
  • ONLINE EXCLUSIVE PRICE
    £54.10
    £52.50 / 2.5 L
    Save £1.60
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-89-5 | CH5N | 31.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-89-5
Molecular Formula CH5N
Molecular Weight (g/mol) 31.06
InChI Key BAVYZALUXZFZLV-UHFFFAOYSA-N
IUPAC Name methanamine
SMILES CN

Specifications

Melting Point -38.0°C
Density 0.9000 g/mL
Boiling Point 48.0°C
Flash Point -15°C
Packaging Glass bottle
Linear Formula CH3NH2
Refractive Index 1.3650 to 1.3700
Quantity 2.5 L
Beilstein 04, 32
Merck Index 15, 6088
Specific Gravity 0.9
Solubility Information Solubility aq. soln.: freely soluble.
Formula Weight 31.06
Grade Extra Pure
Chemical Name or Material Methylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylamine, 40 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.