missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet (Technical Grade), TRC

Product Code. 30135802
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135802 500 mg 500mg
30136895 2.5 g 2.5g
30011508 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135802 Supplier Toronto Research Chemicals Supplier No. TRCM220573500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52080-58-7
Molecular Formula C24 H27 N3
Molecular Weight (g/mol) 357.491
Synonym C.I. Solvent Violet 8, Aizen Methyl Violet Base, Baso Violet 608, Benzenamine, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-, N-Me derivs., Brilliant Oil Violet Base, C.I. 42535:1, C.I. 42535B, Fat Soluble Violet K, Methyl Violet 2B Base
IUPAC Name 4-[[4-(dimethylamino)phenyl]-(4-methyliminocyclohexa-2,5-dien-1-ylidene)methyl]-N,N-dimethylaniline
SMILES CN=C1C=CC(=C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)C=C1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H27N3/c1-25-21-12-6-18(7-13-21)24(19-8-14-22(15-9-19)26(2)3)20-10-16-23(17-11-20)27(4)5/h6-17H,1-5H3
Formula Weight 357.22
Chemical Name or Material Solvent Violet 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.