missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Violet 2B, pure, indicator grade

Product Code. 10125050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
10125050 500 g Glass bottle 500g
10073120 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10125050

Brand: Thermo Scientific Chemicals 228375000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8004-87-3 | C24H28ClN3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H28ClN3
MDL Number MFCD00012568
Synonym Basic Violet 1, C.I. 42535

Specifications

Melting Point 137.0°C
Absorption Ratio 1 to 1.25
Beilstein 13, 755
Solubility Information Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol
Infrared Spectrum Authentic
Lambda Max 580 to 586nm (in 50:50 IMS:water)
Formula Weight 393.95
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
UV_Vis 75% min. (on dry substance)
Color Green
Quantity 500 g
Physical Form Crystalline Powder With Metal Luster
Chemical Name or Material Methyl violet 2B
Show More Show Less
compliance-icons
Product Identifier
  • Methyl Violet 2B, high purity biological stain
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.