missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet 2B, pure, certified

Product Code. 10712464 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10712464 100 g Glass bottle 100g
10249550 25 g Glass bottle 25g
10025443 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10712464 Supplier Thermo Scientific Acros Supplier No. 415071000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8004-87-3 | C24H28ClN3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H28ClN3
MDL Number MFCD00012568
Synonym Basic Violet 1, C.I. 42535

Specifications

Melting Point 137.0°C
Absorption Ratio 1.06 to 1.19
Dye Content 75% min.
Beilstein 13, 755
Solubility Information Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol
Infrared Spectrum Authentic
Lambda Max 583 to 587nm
Formula Weight 393.95
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder With Metal Luster
Chemical Name or Material Methyl Violet 2B, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.